Order Entry
France
Orders LinkContactUsLinkComponent
Calibration oils for kinematic viscometers
Calibration oils for kinematic viscometers
Calibration oils for kinematic viscometers
Calibration oils for kinematic viscometers
The Viscosity standards are designed for calibration of capillary viscometer tubes.
They are packed in 500 ml bottles with the values for the kinematic viscosity at designated temperatures shown on the label. The viscosity value in mm²/s at 20 °C in the table below is indicative and is used for selection of the appropriate standard. The true value is given on the label of each bottle and also on the certificate.
 : Standard specification quoted in mm²/s (cSt), certified in accordance with ASTM D2162
Product Family Options
  • No.
    Viscosity
    Pack type
    Pk
  • S3000
    (25°C) 79747 mm²/s
    Glass bottle
    470 mL
    1 * 470 mL
    1 * 470 mL - 495,00 €
    495,00 €
    1 * 470 mL
       495,00 €
  • S3
    4,6 mm²/s
    Glass bottle
    470 mL
    1 * 470 mL
    1 * 470 mL - 495,00 €
    495,00 €
    1 * 470 mL
       495,00 €
  • S6
    10 mm²/s
    Glass bottle
    470 mL
    1 * 470 mL
    1 * 470 mL - 495,00 €
    495,00 €
    1 * 470 mL
       495,00 €
  • D10
    14 mm²/s
    Glass bottle
    470 mL
    1 * 470 mL
    1 * 470 mL - 495,00 €
    495,00 €
    1 * 470 mL
       495,00 €
  • S20
    43 mm²/s
    Glass bottle
    470 mL
    1 * 470 mL
    1 * 470 mL - 495,00 €
    495,00 €
    1 * 470 mL
       495,00 €
  • S60
    160 mm²/s
    Glass bottle
    470 mL
    1 * 470 mL
    1 * 470 mL - 495,00 €
    495,00 €
    1 * 470 mL
       495,00 €
  • N100
    390 mm²/s
    Glass bottle
    470 mL
    1 * 470 mL
    1 * 470 mL - 495,00 €
    495,00 €
    1 * 470 mL
       495,00 €
  • S200
    650 mm²/s
    Glass bottle
    470 mL
    1 * 470 mL
    1 * 470 mL - 495,00 €
    495,00 €
    1 * 470 mL
       495,00 €
  • S600
    2200 mm²/s
    Glass bottle
    470 mL
    1 * 470 mL
    1 * 470 mL - 495,00 €
    495,00 €
    1 * 470 mL
       495,00 €
  • S2000
    8100 mm²/s
    Glass bottle
    470 mL
    1 * 470 mL
    1 * 470 mL - 495,00 €
    495,00 €
    1 * 470 mL
       495,00 €
  • S8000
    35000 mm²/s
    Glass bottle
    470 mL
    1 * 470 mL
    1 * 470 mL - 495,00 €
    495,00 €
    1 * 470 mL
       495,00 €
  • N35
    79 mm²/s
    Glass bottle
    1
    1 * 1 Pce
    1 * 1 Pce - 495,00 €
    495,00 €
    1 * 1 Pce
       495,00 €