Order Entry
France
ContactUsLinkComponent
Triclosan for synthesis, Sigma-Aldrich®
Triclosan for synthesis, Sigma-Aldrich®
Catalog # 8.16100.0025
Supplier:  Merck
CAS Number:  
Triclosan for synthesis, Sigma-Aldrich®
Catalog # 8.16100.0025
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Solubility:
    0.01 g/l (20 °C)
  • Melting Point:
    55 - 60 °C
  • Molar Mass:
    289.55 g/mol

Specification Test Results

About this item

 :
 :289.54 g/mol
ADR:9,III
 :M-Clarity™ quality level = MQ200
EINECS:222-182-2
UN:3077
 :GHS07
 :GHS09
Storage Temperature:Ambient
CAS Number:3380-34-5
Formula:C₁₂H₇Cl₃O₂
MDL Number:MFCD00800992