Order Entry
France
ContactUsLinkComponent
Triclosan
Triclosan
Catalog # EHERC17803000
Supplier:  EHRENSTORFER
CAS Number:  
Triclosan
Catalog # EHERC17803000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :289.54 g/mol
EINECS:222-182-2
CAS Number:3380-34-5
Formula:C₁₂H₇Cl₃O₂
MDL Number:MFCD00800992