Disodium fumarate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:17013-01-3
:
Formula:C₄H₂Na₂O₄
MDL Number:MFCD00064567
:160.04 g/mol
EINECS:241-087-7
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Titration with HClO4:≥97.5 %
Specification Test Results
Frequently Bought Together