Ammonium-Nitrogen standard solution, 1000 mg/L NH₄ (as N) in water (from (NH₄)Cl) standard for Ion Chromatography
Référence JOBIAS-NH3N9-2Y
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:1000 mg/L
- Pack type:Plastic bottle
- Pk:100 mL
- Description:Ammonium-Nitrogen standard solution
- Contents:NH₄Cl in H₂O
Specifications
About this item
Certifications: Traceable to SRM from NIST, tested in ISO 17034 / ISO 17025 accredited laboratory.