Order Entry
France
ContactUsLinkComponent
Ammonium-Nitrogen standard solution, 1000 mg/L NH₄ (as N) in water
Ammonium-Nitrogen standard solution, 1000 mg/L NH₄ (as N) in water
Référence PRLS88147.260
Supplier:  PROLABO CMP
CAS Number:  
Ammonium-Nitrogen standard solution, 1000 mg/L NH₄ (as N) in water
Référence PRLS88147.260
Supplier:  PROLABO CMP
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Description:
    Ammonium-Nitrogen standard solution

Specifications

About this item