Ammonium-Nitrogen standard solution, 1 mg/L NH₄ (as N) in water
Catalog # PRLSMC313828.0500
Supplier: PROLABO CMP
CAS Number:
Specifications
- Concentration:1 ppm
- Matrix:H₂O
- Pack type:Plastic bottle
- Pk:500 mL
- Description:Ammonium-Nitrogen standard solution