Boron standard solution, 10000 mg/L B in water (from (NH₄)₂B₄O₇.4H₂O) standard for AAS and ICP
634-0144
:
- Concentration:10000 mg/L
- Matrix:H₂O
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Boron
- Symbol:B
: Traceable to SRM from NIST, tested in ISO 17034 / ISO 17025 accredited laboratory.