Bismuth standard solution, 10 mg/L Bi in dil. nitric acid (from Bi) standard for ICP-MS
Catalog # JOBICLBI2-1AY
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:10 mg/L
- Matrix:Nitric acid dil.
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Bismuth
- Symbol:Bi
Specifications
About this item
Certifications: Traceable to SRM from NIST, tested in ISO 17034 / ISO 17025 accredited laboratory.