Antimony standard solution, 10000 mg/L Sb in 1% nitric acid with 6% tartaric acid (from Sb) standard for AAS and ICP
Catalog # JOBIPLSB7-3Y
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:10000 mg/L
- Matrix:H₂O/tr HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Antimony
- Symbol:Sb
Specifications
About this item
Certifications: Traceable to SRM from NIST, tested in ISO 17034 / ISO 17025 accredited laboratory.