Aluminium standard solution, 1000 mg/L Al in 2% hydrochloric acid (from Al) standard for AAS and ICP
Catalog # 634-0132
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:2% HCl
- Pack type:Glass bottle
- Pk:500 mL
- Element:Aluminium
- Symbol:Al
Specifications
About this item
Certifications: Traceable to SRM from NIST, tested in ISO 17034 / ISO 17025 accredited laboratory.