Barium standard solution, 10000 mg/L Ba in 5% nitric acid (from Ba) standard for AAS and ICP
Catalog # 634-0148
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:10000 mg/L
- Matrix:5% HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Barium
- Symbol:Ba
Specifications
About this item
Traceable to SRM from NIST, tested in ISO Guide 34 /ISO17025 accredited laboratory.
Certifications: Traceable to SRM from NIST, tested in ISO 17034 / ISO 17025 accredited laboratory.