Antimony standard solution, 1000 mg/L Sb in 20% hydrochloric acid (from Sb₂O₃) standard for AAS and ICP
Catalog # JOBIPLSB5-2X
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:20% HCl
- Pack type:Plastic bottle
- Pk:500 mL
- Description:Antimony standard solution, 1,000 mg/l Sb in 20% hydrochloric acid (from Sb2O3) standard for AAS and ICP
- Element:Antimony
- Symbol:Sb
Specifications
About this item
Traceable to SRM from NIST, tested in ISO Guide 34 /ISO17025 accredited laboratory.
Certifications: Traceable to SRM from NIST, tested in ISO 17034 / ISO 17025 accredited laboratory.