Beryllium standard solution, 1000 mg/L Be in dil. nitric acid (from Be) standard for ICP-MS
JOBICLBE2-2Y
:
- Concentration:1000 mg/L
- Matrix:Nitric acid dil.
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Beryllium
- Symbol:Be
: Traceable to SRM from NIST, tested in ISO 17034 / ISO 17025 accredited laboratory.