Order Entry
France
ContactUsLinkComponent
Arsenic standard solution, 1,000 mg/L As in dil. nitric acid standard for ICP
Arsenic standard solution, 1,000 mg/L As in dil. nitric acid standard for ICP
  JOBIJYICP-AS
 :  
Arsenic standard solution, 1,000 mg/L As in dil. nitric acid standard for ICP
  JOBIJYICP-AS
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Arsenic
  • Symbol:
    As