Order Entry
France
ContactUsLinkComponent
 
Arsenic standard solution, 1000 mg/L As in dil. nitric acid (from As) standard for ICP-MS
  JOBICLAS2-2Y
 :  
Arsenic standard solution, 1000 mg/L As in dil. nitric acid (from As) standard for ICP-MS
  JOBICLAS2-2Y
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Arsenic
  • Symbol:
    As

 

 

 : Traceable to SRM from NIST, tested in ISO 17034 / ISO 17025 accredited laboratory.