Order Entry
France
ContactUsLinkComponent
Lead standard solution, 1000 mg/L Pb in dil. nitric acid standard for ICP
Lead standard solution, 1000 mg/L Pb in dil. nitric acid standard for ICP
Catalog # JOBIJYICP-PB
CAS Number:  
Lead standard solution, 1000 mg/L Pb in dil. nitric acid standard for ICP
Catalog # JOBIJYICP-PB
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Lead
  • Symbol:
    Pb

Specifications

About this item