Aluminium standard solution, 1000 mg/L Al in dil. nitric acid (from Al) standard for ICP-MS
JOBIJYICP-AL
:
- Concentration:1000 mg/L
- Matrix:2-3% HNO₃
- Pack type:Glass bottle
- Pk:100 mL
- Element:Aluminium
- Symbol:Al
: Traceable to SRM from NIST, tested in ISO 17034 / ISO 17025 accredited laboratory.