Order Entry
France
ContactUsLinkComponent
Nickel sulphate 250 g/l in aqueous solution
Nickel sulphate 250 g/l in aqueous solution
Catalog # PRLS88655.290
Supplier:  PROLABO CMP
CAS Number:  
Nickel sulphate 250 g/l in aqueous solution
Catalog # PRLS88655.290
Supplier:  PROLABO CMP
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Nickel (II) sulphate
 :
 :154.75 g/mol
Storage Temperature:Ambient
EINECS:232-104-9
UN:3082
ADR:9,III
 :GHS08
 :GHS07
 :GHS09
CAS Number:7786-81-4
Formula:NiO₄S
MDL Number:MFCD00011146