Order Entry
France
ContactUsLinkComponent
Nickel sulphate 200 g/l in aqueous solution
Nickel sulphate 200 g/l in aqueous solution
Catalog # PRLS88215.230
Supplier:  PROLABO CMP
CAS Number:  
Nickel sulphate 200 g/l in aqueous solution
Catalog # PRLS88215.230
Supplier:  PROLABO CMP
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Nickel (II) sulphate
 :
 :154.75 g/mol
Storage Temperature:Ambient
EINECS:232-104-9
UN:3082
ADR:9,III
 :GHS08
 :GHS09
CAS Number:7786-81-4
Formula:NiO₄S
MDL Number:MFCD00011146