Order Entry
France
Orders LinkContactUsLinkComponent
Sodium hydroxide 1.5 mol/l (1.5 N) in aqueous solution

Sodium hydroxide 1.5 mol/l (1.5 N) in aqueous solution

Plastic bottle, 500 mL
Catalog # PRLS85685.260
Corrosive
Supplier:   PROLABO CMP
Sodium hydroxide 1.5 mol/l (1.5 N) in aqueous solution
Catalog # PRLS85685.260
Supplier:  PROLABO CMP
CAS Number:  1310-73-2
Sodium hydroxide 1.5 mol/l (1.5 N) in aqueous solution

Plastic bottle, 500 mL

CAS Number:  1310-73-2
Price
39,30 €
Log in for availability
1 * 500 mL
Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Boiling Pt:109 °C (1013 hPa)
  • Density:≤1.11 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-185-5
  • UN:1824
  • ADR:8,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL

Specifications