Tetraoctylphosphonium bromide for synthesis, Sigma-Aldrich®
8.40068.0010
: Merck
:
- Pack type:Plastic bottle
- Pk:10 g
- Seq:3236
- Melting Point:40 - 45 °C
- Molar Mass:563.76 g/mol
- Flash Point:121 °C
- pH:- 3 (50 g/l, H2O)
:
:563.77 g/mol
:8,II
:M-Clarity™ quality level = MQ200
:245-935-7
:3261
:GHS05
:Ambient
:23906-97-0
:C₃₂H₆₈BrP
:MFCD00210101