Order Entry
France
ContactUsLinkComponent
Sodium hydroxide 2.5 mol/l (2.5 N) in aqueous solution
Corrosive
Sodium hydroxide 2.5 mol/l (2.5 N) in aqueous solution
Catalog # PRLS85125.320
Supplier:  PROLABO CMP
CAS Number:  
Corrosive
Sodium hydroxide 2.5 mol/l (2.5 N) in aqueous solution
Catalog # PRLS85125.320
Supplier:  PROLABO CMP
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

About this item

 :
 :40 g/mol
Boiling Pt:109 °C (1013 hPa)
Density:≤1.11 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548