Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Reagent No. 1 for control of chromium (VI )
Reagent No. 1 for control of chromium (VI )
Reagent No. 1 for control of chromium (VI )
PRLS88811.360
:
PROLABO CMP
:
Reagent No. 1 for control of chromium (VI )
PRLS88811.360
:
PROLABO CMP
:
Pk:
5 L
Seq:
5190
Environmentally Preferable:
:
:
Ambient
:
2924
:
3,I
:
GHS02
:
GHS07