Order Entry
France
Orders LinkContactUsLinkComponent
Phenol nitroprussiate solution

Phenol nitroprussiate solution

Glass bottle, 500 mL
Catalog # PRLS88565.260
Supplier:   PROLABO CMP
Phenol nitroprussiate solution
Catalog # PRLS88565.260
Supplier:  PROLABO CMP
CAS Number:  
Phenol nitroprussiate solution

Glass bottle, 500 mL

CAS Number:  
Price
55,70 €
Log in for availability
1 * 500 mL
Product Details & Documents
  • UN:2821
  • ADR:6.1,II

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.