Order Entry
France
Orders LinkContactUsLinkComponent
3,4-Dibromothiophene ≥98%

3,4-Dibromothiophene ≥98%

3,4-Dibromothiophene ≥98%
Certificates
3,4-Dibromothiophene ≥98%
3,4-Dibromothiophene ≥98%

2 Options Available Below

Product Details & Documents

Purity: 98+% CAS No.: 3141-26-2 Molecular Formula: C4H2Br2S Formula Weight: 241.94 Flash Point: 110°(230°F) Refractive Index: 1.6400 Melting Point: 4-5° Boiling Point: 221-222° Beilstein Reference: 107642 MDL No.: MFCD00005465

  • CAS Number:3141-26-2
  • Formula:C₄H₂Br₂S
  • MDL Number:MFCD00005465
  • Molecular Weight:241.93 g/mol
  • Boiling Pt:221…222 °C
  • Melting Pt:4…5 °C
  • Density:2.188
  • Flash Pt:>110 °C (230 °F)
  • Storage Temperature:Ambient
  • EINECS:221-546-8
Product Family Options

Product Information