Order Entry
France
ContactUsLinkComponent
 
2,6-Di-tert-butyl-4-methylpyridine 98%
 
undefined
2,6-Di-tert-butyl-4-methylpyridine 98%

 

  • Appearance:
    Colorless to yellow to orange to light brown Liquid or low melting solid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4760 to 1.4780 (20°C, 589 nm)
  • GC:
    ≥97.5 %

 

 

Frequently Bought Together