Order Entry
France
ContactUsLinkComponent
3,4-Diaminopyridine for synthesis, Sigma-Aldrich®
Toxic
3,4-Diaminopyridine for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Toxic
3,4-Diaminopyridine for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    24 g/l (20 °C)
  • Melting Point:
    217 - 220 °C
  • Molar Mass:
    109.13 g/mol
  • Flash Point:
    240 °C
  • Ignition Point:
    480 °C

Specification Test Results

Related Information

Frequently Bought Together