


1-Naphthylamine for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:134-32-7
:
Formula:C₁₀H₉N
MDL Number:MFCD00004016
:143.19 g/mol
Merck Index:12,06485
Boiling Pt:300 °C (1013 hPa)
Melting Pt:50 °C
Density:1.171 g/cm³ (20 °C)
EINECS:205-138-7
UN:2077
ADR:6.1,III
:M-Clarity™ quality level = MQ200
Flash Pt:156 °C
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Solubility:1.7 g/l (20 °C)
- Melting Point:48 - 50 °C
- Molar Mass:143.19 g/mol
- Bulk Density:560 kg/m3
- Boiling Point:7.1 (1 g/l, H2O, 20 °C)
- Vapor Pressure:301 °C (1013 hPa)
- Flash Point:0.003 hPa (20 °C)
- Refractive Index:157 °C
- Density:1.15 g/cm3 (20 °C)
- Ignition Point:460 °C
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC8.22291.0100_TDS_EN ProductPage_822291
Product specification
msds_822291
SDS
Frequently Bought Together