1-Naphthylamine
EHERC10206350
:
- Pack type:Glass bottle for solids
- Pk:50 mg
- Seq:2370
:
:143.19 g/mol
:12,06485
:300 °C (1013 hPa)
:50 °C
:1.171 g/cm³ (20 °C)
:156 °C
:205-138-7
:2077
:6.1,III
:GHS08
:GHS07
:GHS09
:Ambient
:134-32-7
:C₁₀H₉N
:MFCD00004016