0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
France
ContactUsLinkComponent
Ammonia mercurithiocyanate reagent
Ammonia mercurithiocyanate reagent
Ammonia mercurithiocyanate reagent
Pack type:
Plastic bottle
Pk:
500 mL
Seq:
2725
Environmentally Preferable:
:
:
13,06962
:
130 °C (1013 hPa)
:
40 °C
:
1.04 g/cm³ (20 °C)
:
Ambient
:
244-058-7
:
2471
:
6.1,I
:
GHS06
:
GHS05