Order Entry
France
ContactUsLinkComponent
Silver nitrate ≥99.5%, GPR RECTAPUR®
Oxidizing
Irritant
Corrosive
Environment
Silver nitrate ≥99.5%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Oxidizing
Irritant
Corrosive
Environment
Silver nitrate ≥99.5%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99.5 %
  • Foreign heavy metals (as Pb):
    Max. 20 ppm
  • Not precipitated by HCl (as SO4):
    Max. 100 ppm
  • Cl (Chloride):
    Max. 10 ppm
  • SO4 (Sulphate):
    Max. 100 ppm
  • Fe (Iron):
    Max. 10 ppm
  • Pb (Lead):
    Max. 20 ppm

Specification Test Results

Frequently Bought Together