Order Entry
France
Orders LinkContactUsLinkComponent
 

Silver chloride ≥99%, TECHNICAL

Glass bottle, 25 g
Catalog # 21553.137
Supplier:   VWR Chemicals
Silver chloride ≥99%, TECHNICAL
Catalog # 21553.137
Supplier:  VWR Chemicals
CAS Number:  7783-90-6
Silver chloride ≥99%, TECHNICAL
Silver chloride ≥99%, TECHNICAL

Glass bottle, 25 g

CAS Number:  7783-90-6
Price
404,00 €
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:7783-90-6
  • Formula:AgCl
  • MDL Number:MFCD00003399
  • Molecular Weight:143.32 g/mol
  • Merck Index:13,08582
  • Boiling Pt:1554 °C (1013 hPa)
  • Melting Pt:455 °C
  • Density:5.56 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:232-033-3
Specifications

Specification Test Results

  • Assay:
    Min. 99 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g

Specifications