
Lanthanum(III) oxide ≥99.99% (REO, rare earth oxide basis) for atomic absorption spectroscopy (max. 0.001% Ca)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1312-81-8
:
Formula:La₂O₃
MDL Number:MFCD00011071
:325.81 g/mol
Merck Index:14,5363
Boiling Pt:4200 °C
Melting Pt:2307 °C
Density:6.51
EINECS:215-200-5
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together