Silica gel, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:63231-67-4
:7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232
:60.08 g/mol
Boiling Pt:2230 °C (1013 hPa)
Melting Pt:1710 °C
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
REACH:01-2119379499-16
Frequently Bought Together