Order Entry
France
ContactUsLinkComponent
Silica gel, Supelco®
Silica gel, Supelco®
Catalog # 1.01905.1000
Supplier:  Merck
CAS Number:  
Silica gel, Supelco®
Catalog # 1.01905.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :60.08 g/mol
Boiling Pt:2230 °C (1013 hPa)
Melting Pt:1710 °C
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:231-545-4
REACH:01-2119379499-16
CAS Number:63231-67-4
 :7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232