Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tin (II) chloride
Tin(II) chloride, ultra dry ≥99.9985% (metals basis)
Tin(II) chloride, ultra dry ≥99.9985% (metals basis)
:
Thermo Fisher Scientific
Tin(II) chloride, ultra dry ≥99.9985% (metals basis)
:
Thermo Fisher Scientific
:
7772-99-8
:
:
SnCl₂
:
MFCD00011241
:
189.62 g/mol
:
14,8783
:
652 °C
:
247 °C
:
3.95
:
3260
:
8,III
:
ampouled under argon
:
231-868-0
:
GHS05
:
GHS07
:
Ambient
msds_ALFA035723
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
Methanol ≥99.9% (by GC), LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
: VWR Chemicals
Water USP, JP, Ph. Eur., purified
: VWR Chemicals
Dichloromethane stabilised, EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®
: Merck
Acetone ≥99%, TECHNICAL
: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Acetonitrile ≥99.95%, HiPerSolv CHROMANORM® ULTRA for LC-MS, suitable for UPLC/UHPLC-MS instruments
: VWR Chemicals
2-Propanol, LiChrosolv®, gradient grade for liquid chromatography, Supelco®
: Merck
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
Ethanol absolute ≥99.9% (by GC), EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Ethanol eurodenatured ≥70%, TechniSolv® (1:1:1)
: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals