Order Entry
France
ContactUsLinkComponent
 
Sulphuric acid concentrate 0.1 mol/l (0.2 N) in aqueous solution for ion chromatography, Supelco®
Catalog # 68279-1L
Supplier:  Merck
CAS Number:  
undefined
Sulphuric acid concentrate 0.1 mol/l (0.2 N) in aqueous solution for ion chromatography, Supelco®
Catalog # 68279-1L
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

About this item

 :
 :98.08 g/mol
Boiling Pt:330
Melting Pt:10.38
Density:1.01 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-639-5
UN:2796
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589
ADR:8 II
 :M-Clarity™ quality level = MQ100