Diethyl-3-oxoglutarate
Catalog # MOLEM37576934
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:202.21 g/mol
EINECS:203-302-2
CAS Number:105-50-0
Formula:C₉H₁₄O₅
MDL Number:MFCD00009200