Order Entry
France
ContactUsLinkComponent
Diethyl-3-oxoglutarate
Diethyl-3-oxoglutarate
Catalog # 8.20941.0100
Supplier:  Merck
CAS Number:  
Diethyl-3-oxoglutarate
Catalog # 8.20941.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 ml
  • Seq:
    2440

Specifications

About this item

 :
 :202.21 g/mol
EINECS:203-302-2
CAS Number:105-50-0
Formula:C₉H₁₄O₅
MDL Number:MFCD00009200