Butyl nicotinate
Catalog # 8.00786.0100
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Solubility:2.61 g/l (25 °C)
- Molar Mass:179.22 g/mol
- Boiling Point:122 - 123 °C (11 hPa)
- Vapor Pressure:109 °C
- Flash Point:1.494 (20 °C, 589 nm)
- Density:1.05 g/cm3
Specification Test Results
About this item
:
:179.22 g/mol
:M-Clarity™ quality level = MQ200
EINECS:230-064-7
CAS Number:6938-06-3
Formula:C₁₀H₁₃NO₂
MDL Number:MFCD00006390