Sulphuric acid 95% ≥95%, TECHNICAL
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7664-93-9
:
Formula:H₂SO₄
MDL Number:MFCD00064589
Boiling Pt:330 °C (1013 hPa)
Melting Pt:10.38 °C
Density:1.84 g/cm³ (20 °C)
Molecular Weight:98.08 g/mol
Storage Temperature:Ambient
EINECS:231-639-5
UN:1830
ADR:8 II
REACH:01-2119458838-20
:GHS05
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:Min. 95 %
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Plastic bottle for liquids DIN 45 UN X (ZPACK13962500) 5L Plastic drum DIN 51 UN (ZPACK13951001) 60L Plastic drum for liquids UN (ZPACE8010205)
Packaging Information
spec_VWRC20685.295 spec_VWRC20685.295 spec_VWRC20685.330 spec_VWRC20685.360 spec_VWRC20685.360
Product specification
More
VWR Chemicals Big Packs and Bulks
Product Literature
Frequently Bought Together