Order Entry
France
ContactUsLinkComponent
Sulphuric acid 95-97%, EMPARTA® ACS analytical reagent, Supelco®
Corrosive
Sulphuric acid 95-97%, EMPARTA® ACS analytical reagent, Supelco®
Catalog # 1.01833.2500
Supplier:  Merck
CAS Number:  
undefined
Corrosive
Sulphuric acid 95-97%, EMPARTA® ACS analytical reagent, Supelco®
Catalog # 1.01833.2500
Supplier:  Merck
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble,(caution ! development of heat)
  • Melting Point:
    -20 °C
  • Molar Mass:
    98.08 g/mol
  • Boiling Point:
    335 °C
  • Vapor Pressure:
    0.0001 hPa (20 °C)
  • Density:
    1.84 g/cm3 (20 °C)
  • pH:
    0.3 (49 g/l, H2O, 25 °C)

Specification Test Results

About this item

 :
 :98.08 g/mol
Merck Index:14,08974
Boiling Pt:∼335 °C (1013 hPa)
Melting Pt:3 °C
Density:1.84 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-639-5
UN:1830
ADR:8,II
 :M-Clarity™ quality level = MQ200
 :GHS05
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589