Order Entry
France
ContactUsLinkComponent
Sodium sulphate, anhydrous 98.5-101.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Sodium sulphate, anhydrous 98.5-101.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Sodium sulphate, anhydrous 98.5-101.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (dried substance):
    98.5 - 101.0 %
  • Acidity or alkalinity:
    Passes test
  • Appearance of solution:
    Passes test
  • Identification (SO4):
    Passes test
  • Identification (Na):
    Passes test
  • pH (25°C; 5 %):
    5.2 - 8.0
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Insolubility in water:
    Max. 100 ppm
  • Loss on drying (130°C):
    Max. 0.5 %
  • Loss on ignition (800°C):
    Max. 0.5 %
  • Cl (Chloride):
    Max. 100 ppm
  • PO4 (Phosphate):
    Max. 10 ppm
  • Total N (Nitrogen):
    Max. 5 ppm
  • As (Arsenic):
    Max. 1 ppm
  • Ca (Calcium):
    Max. 50 ppm
  • Fe (Iron):
    Max. 5 ppm
  • K (Potassium):
    Max. 50 ppm
  • Mg (Magnesium):
    Max. 25 ppm
  • Conforms to Reag. Ph.Eur.:
    Passes test

Specification Test Results

Frequently Bought Together