Order Entry
France
ContactUsLinkComponent
Sodium sulphite 95.0-100.5% (by iodometry), EMPROVE® ESSENTIAL Ph. Eur., BP, NF, E221, SAFC®
Sodium sulphite 95.0-100.5% (by iodometry), EMPROVE® ESSENTIAL Ph. Eur., BP, NF, E221, SAFC®
Certificates
Sodium sulphite 95.0-100.5% (by iodometry), EMPROVE® ESSENTIAL Ph. Eur., BP, NF, E221, SAFC®
Certificates

Specification Test Results

  • Assay (Iodometric):
    95.0 - 100.5%
  • Assay (SO₂):
    ≥48.0%
  • Identity:
    Passes Test
  • Appearance of Solution:
    Passes Test
  • Thiosulfate (S₂O₃):
    ≤0.05%
  • Heavy Metals (as Pb):
    ≤0.0005%
  • As (Arsenic):
    ≤0.0002%
  • Hg (Mercury):
    ≤0.0001%
  • Fe (Iron):
    ≤0.0005%
  • Pb (Lead):
    ≤0.0002%
  • Se (Selenium):
    ≤0.00025%
  • Zn (Zinc):
    ≤0.0025%
  • Residual solvents (Ph. Eur./USP/ICH):
    Excluded by the Manufacturing Process

Specification Test Results

Related Information

Frequently Bought Together