Order Entry
France
Orders LinkContactUsLinkComponent
Sodium hydroxide

Sodium hydroxide

Glass bottle for solids, 115 g
Catalog # PRLS87256.190
Corrosive
Supplier:   PROLABO CMP
Sodium hydroxide
Catalog # PRLS87256.190
Supplier:  PROLABO CMP
CAS Number:  1310-73-2
Sodium hydroxide

Glass bottle for solids, 115 g

CAS Number:  1310-73-2
Price
21,50 €
Log in for availability
1 * 115 g
Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Merck Index:14,08627
  • Boiling Pt:1390 °C (1013 hPa)
  • Melting Pt:323 °C
  • Density:2.13 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-185-5
  • UN:1823
  • ADR:8,II
  • REACH:01-2119457892-27

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    115 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.