Order Entry
France
ContactUsLinkComponent
 
 
Sodium iodide 99.0-100.5% Ph. Eur.
 
undefined
 
Sodium iodide 99.0-100.5% Ph. Eur.
 

 

  • Assay (calculated on dried substance):
    99.0 - 100.5 %
  • Appearance:
    Conforms (see CoA/CoS)
  • Identification A:
    Passes test
  • Identification B:
    Passes test
  • Appearance of solution:
    Passes test
  • Alkalinity:
    Passes test
  • IO3 (Iodate):
    Passes test
  • SO4 (Sulphate):
    Max. 150 ppm
  • S2O3 (Thiosulphate):
    Passes test
  • Fe (Iron):
    Max. 20 ppm
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Loss on drying (105°C):
    Max. 3 %
  • Residual solvents:
    Passes test

 

Frequently Bought Together