Sodium iodide ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7681-82-5
:
Formula:NaI
MDL Number:MFCD00003532
:149.89 g/mol
Merck Index:13,08705
Boiling Pt:1300 °C (1013 hPa)
Melting Pt:662 °C
Density:3.67 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-679-3
UN:3077
ADR:9,III
:GHS09
Specification Test Results
- Assay:Min. 99 %
- Heavy metals (as Pb):Max. 10 ppm
- Cl + Br (as Cl):Max. 0.05 %
- SO4 (Sulphate):Max. 0.015 %
- Fe (Iron):Max. 10 ppm
Specification Test Results
Frequently Bought Together