Order Entry
France
ContactUsLinkComponent
Sodium iodide, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, SAFC®
Sodium iodide, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, SAFC®
Catalog # 1.06520.5000
CAS Number:  
Sodium iodide, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, SAFC®
Catalog # 1.06520.5000
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    5 kg
  • Seq:
    4114

Specifications

About this item

 :
 :149.89 g/mol
Merck Index:13,08705
Boiling Pt:1300 °C (1013 hPa)
Melting Pt:662 °C
Density:3.67 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-679-3
UN:3077
ADR:9,III
 :GHS08
 :GHS09
CAS Number:7681-82-5
Formula:NaI
MDL Number:MFCD00003532