Order Entry
France
ContactUsLinkComponent
 
Sodium iodide, Suprapur® for trace analysis, Supelco®
  1.06519.0100
 :  Merck
 :  
Sodium iodide, Suprapur® for trace analysis, Supelco®
  1.06519.0100
 :  Merck
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

 

 

  • Solubility:
    1793 g/l (20 °C)
  • Melting Point:
    662 °C
  • Molar Mass:
    149.89 g/mol
  • Bulk Density:
    1500 - 2000 kg/m3
  • Boiling Point:
    1304 °C (1013 hPa)
  • Vapor Pressure:
    1.3 hPa (767 °C)
  • Density:
    3.67 g/cm3 (20 °C)
  • pH:
    6 - 9 (50 g/l, H2O, 20 °C)
  • Hazard Class:
    9
  • Hazard Class Description:
    Miscellaneous Hazardous Material

 

 

 :
 :149.89 g/mol
 :13,08705
 :1300 °C (1013 hPa)
 :662 °C
 :3.67 g/cm³ (20 °C)
 :Ambient
 :231-679-3
 :3077
 :9,III
 :M-Clarity™ quality level = MQ100
 :GHS08
 :GHS09
 :7681-82-5
 :NaI
 :MFCD00003532